Learn More
Memmert Signal cable 3m for pump control (not required with pump module), for VO29
Brand:  Memmert VO29B39410
Signal cable 3m for pump control (not required with pump module), for VO29
Product Code. 16374537
  • ONLINE EXCLUSIVE PRICE
    196.00€
    189.00€ / Each
    Save 7.00€
    3% Off
Only null left
Estimated Shipment: 10-03-2026