Learn More
RETSCH Sieve Shaker AS 200 Digital
Product Code. 10045120 Shop All Retsch Products
Click to view available options
Load Bearing Capacity (Metric):
3 kg
Unit Size:
Each
Product Code. 10045120
Brand: RETSCH 30.015.0001
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Specifications
Load Bearing Capacity (Metric) 3 kg
Electrical Requirements 230 V, 50 Hz
Voltage 230 V
Product Type Sieve Shaker
Frequency 50 Hz
Depth (Metric) 350 mm
Height (Metric) 230 mm