Learn More
BOLA SICCO Desiccant
Product Code. 17197714
Change view
Click to view available options
Product Type:
Desiccator Canister
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
17197714 Desiccator Canister Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17197714 Supplier BOLA Supplier No. V 189508
  • ONLINE EXCLUSIVE PRICE
    62.68€
    48.49€ / Pack of 1
    Save 14.19€
    22% Off
 Restricted Item
Estimated Shipment: 07-12-2026
High-quality silica gel desiccant specifically designed for use in their desiccator cabinets, easy regeneration at approx. 90 - 110°C
  • Silica gel with colour indicator from orange to dark brown and black
  • Does not contain cobalt-II-chloride
  • Grain size 2 - 5 mm, loss after drying less or equal to 4%, Weight 720 g
Specifications
Material Silica Gel
Weight (Metric) 720 g
Product Type Desiccator Canister