Learn More
Afora™ Short Neck Round Flask
Product Code. p-8015396
Click to view available options
Height (Metric):
73 mm
88 mm
100 mm
113 mm
152 mm
182 mm
217 mm
Unit Size:
Each
Product Code. 12090362
Brand: Afora™ 8/29E
Only null left
Add to basket
Add to basket
Boil laboratory chemicals using the Afora™ Short Neck Round Flask. Its quality construction makes it durable and reliable in the laboratory.
  • Glass
  • Spherical bottom
  • Shorter neck
Specifications
Capacity (Metric) 500 mL
Material Borosilicate Glass
Height (Metric) 152 mm
Neck Style Single
Bottom Shape Round