Learn More
Afora™ Short Neck Round Flask
Product Code. 11996608
Click to view available options
Height (Metric):
73 mm
88 mm
100 mm
113 mm
152 mm
182 mm
217 mm
Unit Size:
Each
Product Code. 11996608
Brand: Afora™ 3/29E
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Boil laboratory chemicals using the Afora™ Short Neck Round Flask. Its quality construction makes it durable and reliable in the laboratory.
  • Glass
  • Spherical bottom
  • Shorter neck
Specifications
Capacity (Metric) 50 mL
Material Borosilicate Glass
Height (Metric) 100 mm
Neck Style Single
Bottom Shape Round