Learn More
Afora™ Short Neck Round Flask
Boil laboratory chemicals using the Afora™ Short Neck Round Flask. Its quality construction makes it durable and reliable in the laboratory.
Brand: Afora™ 2/E
Additional Details : Weight : 0.18900kg
Description
- Glass
- Spherical bottom
- Shorter neck
Specifications
25 mL | |
73 mm | |
Round |
Borosilicate Glass | |
Single |