Learn More
Afora™ Short Neck Round Flask
Click to view available options
Height (Metric):
73 mm
88 mm
100 mm
113 mm
152 mm
182 mm
217 mm
Unit Size:
Each
Description
- Glass
- Spherical bottom
- Shorter neck
Specifications
Specifications
| Capacity (Metric) | 2000 mL |
| Material | Borosilicate Glass |
| Height (Metric) | 217 mm |
| Neck Style | Single |
| Bottom Shape | Round |