Learn More
SHKBP1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA521731
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: A431, H1299. Predicted reactivity: Mouse (96%), Rat (96%), Xenopus laevis (82%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Inhibits CBL-SH3KBP1 complex mediated down-regulation of EGFR signaling by sequestration of SH3KBP1. Binds to SH3KBP1 and prevents its interaction with CBL and inhibits translocation of SH3KBP1 to EGFR containing vesicles upon EGF stimulation.Specifications
SHKBP1 | |
Polyclonal | |
Unconjugated | |
SHKBP1 | |
B930062H15Rik; PP203; RGD1309281; Sb1; SETA binding protein 1; SETA-binding protein 1; SH3KBP1 binding protein 1; SH3KBP1-binding protein 1; Shkbp1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
92799 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q8TBC3 | |
SHKBP1 | |
Recombinant fragment corresponding to a region within amino acids 162 and 477 of SHKBP1 (Uniprot ID#Q8TBC3) | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |