Learn More
Ohaus Shipping Scale Courier 5000 i-C52M50R EU 50kg/0.02kg
Brand:  Ohaus 30745892
Shipping Scale Courier 5000 i-C52M50R EU 50kg/0.02kg
Product Code. 17911382
  • ONLINE EXCLUSIVE PRICE
    334.00€
    325.00€ / Each
    Save 9.00€
    2% Off