Learn More
Hospidex™ Sharpsafe™ Polypropylene Biohazard Container
Product Code. 11992089
Change view
Click to view available options
Capacity (Metric):
1 L
2 L
7 L
13 L
Unit Size:
Pack of 10
Pack of 100
Pack of 20
Pack of 50
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
11992089 7 L Pack of 10
11905386 13 L Pack of 20
11988877 1 L Pack of 100
11972089 1 L Pack of 20
11994106 2 L Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11992089 Supplier Hospidex™ Supplier No. SHA41105610
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Use the Sharpsafe™ Polypropylene Biohazard Container to transport dangerous biohazardous goods for disposal.
  • Yellow
  • Polypropylene
  • NFX 30-505 certification
Specifications
Capacity (Metric) 7 L
Certifications/Compliance NFX30-500
Height (Metric) 188 mm
Length (Metric) 106 mm
Autoclavable Autoclavable
For Use With (Application) For collection of sharps waste treatment activities, pungent, sharp and / or pointed
Material Polypropylene
Product Type Biohazard Container