Learn More
Hospidex™ SharpSafe™ Biohazard Carton Bags
Product Code. p-8009819
Click to view available options
Capacity (Metric):
20 L
50 L
Color:
Brown
White
Unit Size:
Pack of 10
Product Code. 11928887
Brand: Hospidex™ ONDANDEC50
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Use SharpSafe™ Biohazard Carton Bags for effective waste disposal. These containers facilitate the process that is critical not only to avoid regulatory fines but also to prevent the spread of infectious diseases.
  • Cardboard
  • Various sizes and colors
  • Disposable
Specifications
Capacity (Metric) 50 L
Color Brown
Disposable Single-use
Material Cardboard
Length (Metric) 50 mm
Product Type Biohazard Carton Bag