Learn More
Sharpsafe™ Impenetrable Polypropylene Disposal Containers
Product Code. p-8010566
Change view
Click to view available options
Capacity (Metric):
30 L
60 L
Unit Size:
Each
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
11879550 60 L Each
11869550 30 L Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11879550 Supplier Sharpsafe™ Supplier No. SANBG60LADTEGH+
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use Sharpsafe™ Impenetrable Polypropylene Disposal Containers for incinerations or alternative treatments and for outstanding, impenetrable protection from leaks or sharps.
  • Yellow with orange lid
  • Clear British Standard markings
  • Visible fill line
Specifications
Capacity (Metric) 60 L
Certifications/Compliance Manufactured to BS 7320: 1990, ISO 9002 standards and UN Packing Group II approved
Closure Type White Lid
Length (Metric) 60 mm
Model Rectangular
Quantity 1 Ea.
Includes Closure
Material Polypropylene
Product Type Rectangular Container