Learn More
Sharpsafe™ Impenetrable Polypropylene Disposal Containers
Click to view available options
Capacity (Metric):
30 L
60 L
missing translation for 'unitSize'
Each
Pack of 10
Description
- Yellow with orange lid
- Clear British Standard markings
- Visible fill line
Specifications
Specifications
| Capacity (Metric) | 30 L |
| Certifications/Compliance | Manufactured to BS 7320: 1990, ISO 9002 standards and UN Packing Group II approved |
| Closure Type | Orange Lid |
| Model | Rectangular |
| Quantity | 1 Ea. |
| Includes | Closure |
| Material | Polypropylene |
| Product Type | Container |