Learn More
Honeywell™ SHARPFLEX PU Gloves
Click to view available options
Size:
7
8
9
10
11
Unit Size:
1 pair
Description
- Knitted glove in composite and para-aramid yarn
- The knitting technique provides the glove with excellent fit and high dexterity
- The composite yarn is reliable and safe protection
- The para-aramid yarn assure a resistance to cut risks
- Plated polyamide inside
- Offer a greater comfort and limit the contact of the skin with composite fiber
- Black PU coating on palm
- Increase the abrasion resistance and allows to use in slighy greasy environements
Specifications
Specifications
| Certifications/Compliance | EN 420-03 EEC Basic standards, EN 388-03 Mechanical hazards |
| Description | Para-aramid and composite fiber, 13 gauge |
| Coating Material | Polyurethane |
| Color | Green, Yellow, Black |
| Size | 8 |
| Length (Metric) | 25 cm |