Learn More
Honeywell™ SHARPFLEX NIT Gloves
Product Code. p-7159532
Click to view available options
Size:
7
8
9
10
11
Unit Size:
1 pair
Product Code. 15876735
Brand: Honeywell™ 223252407
Edge
Honeywell™ SHARPFLEX NIT Gloves for handling heavy parts with high cut risks in a dry or slightly greasy environment.
  • Knitted glove in composite and para-aramid yarn
  • The knitting technique provides the glove with excellent fit and high dexterity
  • The composite yarn is reliable and safe protection
  • The para-aramid yarn assure a resistance to the contact heat and cut risks
  • Plated polyamide inside offers a greater comfort and limit the contact of the skin with composite fiber
  • Black nitrile foam coating on palm
  • Nitrile coating provides good protection against oil and grease
  • The foam coating makes the glove more flexible than standard nitrile
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN 420-03 EEC Basic standards, EN 388-16 Mechanical hazards
Description Para-aramid and composite, 13 gauge
Color Green, Yellow, Black
Material Nitrile, Polyamide
Size 7
Length (Metric) 25 cm