Learn More
Honeywell™ SHARPFLEX Latex Gloves
Click to view available options
Size:
7
8
9
10
Unit Size:
1 pair
Description
- Knitted glove in composite and para-aramid yarn
- The knitting technique provides the glove with excellent fit and high dexterity
- The composite yarn is reliable and safe protection
- The para-aramid yarn assure a resistance to the contact heat and cut risks
- Plated polyamide inside
- Offer a greater comfort and limit the contact of the skin with composite fiber
- Blue crinkled latex coating on palm
- Latex coating provides better grip and a light thermal protection (up to 250°C) resistance
Specifications
Specifications
| Certifications/Compliance | EN 420-03 EEC Basic standards, EN 388-03 Mechanical hazards, EN407-04 : Thermal risks |
| Description | Para-aramid and composite, 13 gauge |
| Color | Yellow, Green, Blue |
| Material | Latex |
| Size | 7 |
| Length (Metric) | 25 cm |