Learn More
Epredia™ Embedding Tissue Cassettes
Product Code. 15782482 Shop All Epredia Products
Click to view available options
Color:
Blue
Unit Size:
Each
Product Code. 15782482
Brand: Epredia™ B1003500BL
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Hold and identify tissue samples in processing, embedding, and sectioning procedures with Epredia™ Embedding Tissue Cassettes.
  • Designed to accept metal lids
  • Designed with round hole pores
  • Supplied in boxes of 500 cassettes
TRUSTED_SUSTAINABILITY
Specifications
Pore Style Round Holes
Color Blue
Product Type Tissue Cassette