Learn More
Epredia™ Shandon™ Stainless-Steel Scalpel Blade Handle, Holds No. 20-25 Blades
Product Code. 12697746 Shop All Epredia Products
Change view
Click to view available options
Size:
# 4
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
12697746 # 4 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12697746 Supplier Epredia™ Supplier No. 5334
  • ONLINE EXCLUSIVE PRICE
    58.05€
    49.87€ / Pack of 1
    Save 8.18€
    14% Off
Add to basket
Edge
Add to basket
Securely hold blades with the Epredia™ Shandon™ Stainless-Steel Scalpel Blade Handle, constructed of #4 stainless steel.
  • Properly hardened, tempered and passivated
  • Electronically polished and cleaned for a high-quality finish
  • Quality patterns, dimensions and finish held to exacting standards, producing the finest instruments available
  • Defect-free in material and workmanship
TRUSTED_SUSTAINABILITY
Specifications
Blade Material Stainless Steel
Size # 4
Length (English) 8.5 in.
Length (Metric) 21.6 cm
For Use With (Equipment) Holds # 20 to # 25 Blade