Learn More
Epredia™ Shandon™ Russian Thumb Forceps, Circular Serrated Tips, standard, 8 in. (20.3cm)
Product Code. p-4520658 Shop All Epredia Products
Click to view available options
Grade:
Standard
Length (Metric):
15.2 cm
Unit Size:
1 pair
Product Code. 15246219
Brand: Epredia™ 6551
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Epredia™ Shandon Russian Thumb Forceps are available in premium and standard grades.
Features:
  • Electronically polished and cleaned for a high-quality finish
  • Patterns, dimensions and finish held to exacting standards
  • Guaranteed to be free from defects in material and workmanship
  • Available in three lengths
  • Come in premium or standard grades
TRUSTED_SUSTAINABILITY
Specifications
Product Type Russian Thumb Forceps
Grade Standard
Length (English) 6 in.
Length (Metric) 15.2 cm