Learn More
Epredia™ Shandon™ Plastic Scalpel Blade Handle, Holds No. 20-25 Blades
Product Code. 12687896 Shop All Epredia Products
Change view
Click to view available options
For Use With (Equipment):
# 20 to # 25 Blade for Deep Surgery
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
12687896 # 20 to # 25 Blade for Deep Surgery Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12687896 Supplier Epredia™ Supplier No. 3120027
  • ONLINE EXCLUSIVE PRICE
    64.30€
    55.28€ / Pack of 1
    Save 9.02€
    14% Off
Estimated Shipment: 16-12-2026
Securely hold blades with the autoclavable Epredia™ Shandon™ Plastic Scalpel Blade Handle, constructed of red plastic #6.
  • Properly hardened, tempered and passivated
  • Electronically polished and cleaned for a high-quality finish
  • Quality patterns, dimensions and finish held to exacting standards, producing the finest instruments available
  • Defect-free in material and workmanship
TRUSTED_SUSTAINABILITY
Specifications
Blade Material Plastic
For Use With (Equipment) # 20 to # 25 Blade for Deep Surgery