Learn More
Epredia™ Shandon™ Plain Bone Shears, Screw Pivot, 1 Spring, standard, 9 in. (22.9cm)
Product Code. 15664444 Shop All Epredia Products
Change view
Click to view available options
Length (English):
9 in.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (English) Unit Size
15664444 9 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15664444 Supplier Epredia™ Supplier No. 50619
  • ONLINE EXCLUSIVE PRICE
    183.00€
    156.16€ / Pack of 1
    Save 26.84€
    14% Off
 Restricted Item
In Stock
Make precise cuts through tough bone tissue with Epredia™ Shandon™ Plain Bone Shears, featuring a single spring and a screw-pivot.
Consistent Quality
  • Properly hardened, tempered and passivated
  • Electronically polished and cleaned for a high-quality finish
  • Patterns, dimensions and finish held to exacting standards
  • Guaranteed to be free from defects in material and workmanship
TRUSTED_SUSTAINABILITY
Specifications
Product Type Plain Bone Shears
Grade Standard
Length (English) 9 in.
Length (Metric) 22.9 cm