Learn More
Epredia™ Peel-A-Way™ Disposable Embedding Molds
Product Code. p-4524719 Shop All Epredia Products
Click to view available options
Dimensions (L x W x H):
22 x 22 x 20 mm
22 x 30 x 20 mm
22 x 40 x 20 mm
22x22 to 12x12 mm, 20 mm deep
Shape:
Rectangular
Square
Truncated, tapered
Unit Size:
Each
Pack of 264
Pack of 288
Product Code. 12677736
Brand: Epredia™ 1830
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Promote rapid paraffin hardening with Epredia Peel-A-Way Disposable Embedding Molds.
  • Constructed of rigid transparent plastic.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Embedding Mold
Material Plastic
Dimensions (L x W x H) 22 x 30 x 20 mm
Disposable Single-use
Shape Rectangular