Learn More
Epredia™ Shandon™ Octospot™ Accessories
Product Code. 12510057 Shop All Epredia Products
Click to view available options
Type:
8-well Strips
8-well Strips, Loose
EZ-Write Slides, 8 spots
Holding Block
Solvent-Resistant Slides, 8 spots
Thick Filter Cards
Unit Size:
Each
Pack of 100
Pack of 5
Pack of 60
Product Code. 12510057
Brand: Epredia™ 1001646
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
These Epredia™ accessories are for use with Shandon Octospot Starter Packs.
TRUSTED_SUSTAINABILITY
Specifications
Type EZ-Write Slides, 8 spots
For Use With (Equipment) Shandon™ Octospot™ Starter Packs