Learn More
Epredia™ Shandon™ Microdissecting Extra Fine Tip Thumb Forceps, Extra Fine Tips, curved, 4.5 in. (11.4cm)
Product Code. 15327758 Shop All Epredia Products
Change view
Click to view available options
Length (English):
4.5 in.
Unit Size:
1 pair
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (English) Unit Size
15327758 4.5 in. 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15327758 Supplier Epredia™ Supplier No. 3120018
  • ONLINE EXCLUSIVE PRICE
    161.00€
    137.35€ / 1 pairs
    Save 23.65€
    14% Off
Choose 4.5 in. (11.4cm) microdissecting extra fine tip thumb forceps with extra fine tips and curved arms for quality and precision.
Features:
  • Nonmagnetic stainless-steel
  • Properly hardened, tempered and passivated
  • Electronically polished and cleaned for a high-quality finish
  • Patterns, dimensions and finish held to exacting standards
  • Guaranteed to be free from defects in material and workmanship
TRUSTED_SUSTAINABILITY
Specifications
Length (English) 4.5 in.