Learn More
Epredia™ Accessories for Hyperclean TruAir™ Workstation
For use with Epredia™ Hyperclean Pathology Workstations.
158.33€ - 245.00€
Product Code | Brand | Description | For Use With (Equipment) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | For Use With (Equipment) | Price | Quantity & Availability | ||||
12623106
|
Epredia™
9990610 |
Size: 29 cm x 15 cm x 3.3 cm | Epredia Hyperclean Pathology Workstations, Models 1, 20, 22, 36, 60 |
183.00€
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
||||
12613106
|
Epredia™
9990612 |
Size: 28.7 cm x 14.7 cm x 3.5 cm | Epredia Hyperclean Pathology Workstations |
245.00€
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
||||
10443210
|
Epredia™
9990612 |
Size: 28.7 cm x 14.7 cm x 3.5 cm | Epredia Hyperclean Pathology Workstations |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||||
Description
Accessories:- Charcoal Filter
- Fine particle Filters
- Formaldehyde Neutralizing (Potassium Permanganate) Filter
Specifications
Hyperclean™ |