Learn More
Epredia™ Glyo-Fixx™
Obtain superior results to neutral buffered formalin with Glyo-Fixx™, a noncarcinogenic, nonsensitizing, and environmentally friendly substitute.
74.00€ - 273.00€
Product Code | Brand | Product Type | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Quantity | Price | Quantity & Availability | ||||
10249908
|
Epredia™
6764260 |
Fixative | 5 gal. |
251.00€
Each |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||||
10602689
|
Epredia™
6764262 |
Fixative | 1 gal. |
74.00€
3.80 L |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||||
12520067
|
Epredia™
9990928 |
Fixative | 125 x 120mL |
273.00€
Pack of 125 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||||
12500067
|
Epredia™
9990924 |
Fixative | 125 x 60mL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||||
Description
- Produces distinct and crisp cellular morphology
- Fixes most antigens without impairing immunoreactivity
- Allows special stain dyes, including silver procedures, to be sharp without background staining
- Presents fine detail in specimen structure
Specifications
Fixative |