Learn More
Epredia™ Shandon™ Fine Point Dissecting Scissors, Angular, Premium, 4.5 in. (11.4cm)
Product Code. 15644444 Shop All Epredia Products
Click to view available options
Grade:
Premium
Standard
Unit Size:
Each
Product Code. 15644444
Brand: Epredia™ 11999
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Epredia™ Shandon Fine Point Dissecting Scissors are available in Premium and Standard grades.
  • Properly hardened, tempered and passivated
  • Electronically polished and cleaned for a high-quality finish
  • Patterns, dimensions and finish held to exacting standards, producing the finest instruments available
  • Guaranteed to be free from defects in material and workmanship
TRUSTED_SUSTAINABILITY
Specifications
Product Type Dissecting Scissors
Blade Style Angled
Grade Standard
Length (English) 4.5 in.
Length (Metric) 11.4 cm
Tip Style Fine Point