Learn More
Epredia™ EZ Double Cytofunnel™
Double the number of sample presentations per run with the Epredia™ EZ Double Cytofunnel™.
825.00€ - 1250.00€
Specifications
Color | White |
---|---|
Disposable | Single-use |
Type | EZ Double Cytofunnel™, With Filter Cards and Caps |
Volume (Metric) | 0.05 to 0.25 mL |
Product Code | Brand | Quantity | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantity | Prezzo | Quantità e disponibilità | |||||
11911788
|
Epredia™
A78710005 |
40 Pk. |
825.00€
Pack of 40 |
Estimated Shipment: 19-08-2024
Log in to see stock. |
|||||
12076689
|
Epredia™
A78710021 |
200/Cs. |
1250.00€
Pack of 200 |
Estimated Shipment: 19-08-2024
Log in to see stock. |
|||||
Descrizione
- Disposable sample chambers with white filter cards and caps
- Funnel markings corresponding to slide markings for simple identification
- Pre-attached filter card
Specifica
White | |
Single-use | |
EZ Double Cytofunnel™, With Filter Cards and Caps | |
0.05 to 0.25 mL |