Learn More
Epredia™ Shandon™ Embedding Rings
Product Code. p-4524751 Shop All Epredia Products
Click to view available options
Color:
Blue
Unit Size:
Pack of 250
Product Code. 12540077
Brand: Epredia™ 1000956
Identify samples in storage with Epredia™ Shandon™ Embedding Rings, which securely hold tissue samples for sectioning.
  • Made of high impact polystyrene
  • Fits well in microtome chuck adapters
  • Supplied in boxes of 250 rings
TRUSTED_SUSTAINABILITY
Specifications
Color Blue
Material Polystyrene
Product Type Embedding Rings