Learn More
Epredia™ Embedding Cassettes in Tube Packs
Product Code. 15145944 Shop All Epredia Products
Click to view available options
Color:
Aqua
Blue
Gray
Green
Lilac
Orange
Peach
Pink
Tan
White
Yellow
Unit Size:
Each
Pack of 750
Product Code. 15145944
Brand: Epredia™ B1000732YW
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Hold and identify tissue samples in processing, embedding and sectioning procedures with Embedding Cassettes in Tube Packs. Compatible for use with the Epredia™ PrintMate AS™ Cassette Printer.
  • Can be used on all Epredia PrintMate automatic cassette labelers
  • Fit securely in microtome chuck adapters
  • Designed to accept metal lids (Order number 1001003, 25/pk., sold separately)
  • Made of acetal polymer with a pore style of round holes
  • Supplied in 10 tubes of 75 cassettes each
TRUSTED_SUSTAINABILITY
Specifications
Pore Style Round Holes
Quantity 750 Cassettes/Cs.
Color Yellow
Material Acetal Polymer