Learn More
Epredia™ Embedding Cassettes in Tube Packs
Shop All Epredia Products
Click to view available options
Color:
Aqua
Blue
Gray
Green
Lilac
Orange
Peach
Pink
Tan
White
Yellow
Unit Size:
Each
Pack of 750
Description
- Can be used on all Epredia PrintMate automatic cassette labelers
- Fit securely in microtome chuck adapters
- Designed to accept metal lids (Order number 1001003, 25/pk., sold separately)
- Made of acetal polymer with a pore style of round holes
- Supplied in 10 tubes of 75 cassettes each
Specifications
Specifications
| Pore Style | Round Holes |
| Quantity | 750 Cassettes/Cs. |
| Color | Green |
| Material | Acetal Polymer |