Learn More
Epredia™ Shandon™ Embedding Cassettes II in Tube Packs, aqua
Shop All Thermo Scientific Products
Click to view available options
Color:
Aqua
Blue
Gray
Green
Lilac
Orange
Peach
Pink
Tan
White
Yellow
Unit Size:
Pack of 750
Description
- Acetal polymer cassettes with writing face at 45° angle
- Slotted pore style
- Locking lid attached by two hinges
Ordering Information:
- Tube packs are supplied in 10 tubes of 75 cassettes each
Specifications
Specifications
| Pore Style | Slotted Pore |
| Writing Face Angle | 45° |
| Color | Lilac |
| Material | Acetal Polymer |
| Product Type | Biopsy Cassette |