Learn More
Epredia™ Shandon™ Dissecting Tray for Raised Dissecting Boards, 12x 11 in.
Product Code. 12663026 Shop All Thermo Scientific Products
Change view
Click to view available options
Length (English):
23 in.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (English) Unit Size
12663026 23 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12663026 Supplier Epredia™ Supplier No. 3611403
  • ONLINE EXCLUSIVE PRICE
    532.00€
    452.93€ / Pack of 1
    Save 79.07€
    14% Off
 Restricted Item
In Stock
Epredia™ Shandon Dissecting Tray for Raised Dissecting Boards feature ample space for drainage and instruments.
  • Four embossments on tray bottom hold Raised Dissecting Boards 356 and 35614
TRUSTED_SUSTAINABILITY
Specifications
Product Type Dissecting Tray
Material Polyethylene
Size Large
Length (English) 23 in.
Width (English) 16 in.