Learn More
Epredia™ Shandon™ Disposable Base Molds, 0.95 x 0.95 x 0.2 in. (24 x 24 x 5mm)
Product Code. 12559036 Shop All Epredia Products
Click to view available options
Length (Metric):
7 mm
15 mm
24 mm
Width (Metric):
7 mm
15 mm
24 mm
30 mm
37 mm
Unit Size:
Pack of 500
Product Code. 12559036
Brand: Epredia™ 41741
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Promote rapid paraffin hardening with Epredia™ Shandon™ Disposable Base Molds.
  • Constructed of rigid transparent plastic
TRUSTED_SUSTAINABILITY
Specifications
Product Type Base Mold
Material Plastic
Length (Metric) 15 mm
Width (Metric) 15 mm
Length (English) 0.59 in.
Width (English) 0.59 in.
Depth (Metric) 5 mm
Depth (English) 0.2 in.