Learn More
Epredia™ Cytospin™ Collection Fluid
Product Code. 12570057 Shop All Epredia Products
Click to view available options
Quantity:
120mL
500mL
4 L
Unit Size:
15 L
3.8 L
Pack of 125
Pack of 2
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 12570057
Brand: Epredia™ 9990323
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Store specimens or unstained slides for weeks without adverse effect with Epredia™ Shandon™ Cytospin™ Collection Fluid.
  • Packaged in convenient pump bottle or pre-filled specimen cups
  • Ready-to-use alcohol/Carbowax™ (PEG) combination for direct specimen collection and fixation
TRUSTED_SUSTAINABILITY
Specifications
Product Type Collection Fluid
Quantity 120mL
Solution Type Alcohol/Carbowax (PEG) combination