Learn More
Epredia™ Shandon™ Broad Point Dressing Thumb Forceps, Fine serrated tip, premium, 5.5 in. (14.0cm)
Product Code. 15674444 Shop All Epredia Products
Change view
Click to view available options
Grade:
Premium
Standard
Length (English):
4.5 in.
5 in.
5.5 in.
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Grade Length (English) Unit Size
15674444 Standard 5.5 in. Each
15617175 Premium 4.5 in. Each
15614454 Premium 5 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15674444 Supplier Epredia™ Supplier No. 6303
  • ONLINE EXCLUSIVE PRICE
    21.70€
    18.64€ / Pack of 1
    Save 3.06€
    14% Off
 Restricted Item
In Stock
Select a fine serrated tip ranging in size from 4.5 to 12 in. (11.4 to 30.5cm) on the Shandon™ Broad Point Dressing Thumb Forceps.
Features:
  • Properly hardened, tempered and passivated
  • Electronically polished and cleaned for a high-quality finish
  • Patterns, dimensions and finish held to exacting standards
TRUSTED_SUSTAINABILITY
Specifications
Product Type Thumb Forceps
Grade Standard
Length (English) 5.5 in.
Length (Metric) 14 cm