Learn More
Epredia™ Shandon™ Biopsy Processing/Embedding Cassettes II in Tube Packs, aqua
Product Code. 12973765 Shop All Epredia Products
Click to view available options
Color:
Aqua
Blue
Fluorescent Green
Fluorescent Pink
Fluorescent Yellow
Gray
Green
Lilac
Orange
Peach
Pink
Tan
White
Yellow
Unit Size:
Each
Pack of 750
Product Code. 12973765
Brand: Epredia™ B1000730FYW
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Aid the process of speedy and reliable biopsy specimen labeling with Epredia™ Shandon™ Biopsy Processing/Embedding Cassettes II in Tube Packs.
  • Writing face is flat 45° angle
  • Locking lid attached by two hinges
  • Front or back opening
Ordering Information:
  • Packaged in 75-cassette tubes
TRUSTED_SUSTAINABILITY
Specifications
Pore Style Small Square Pore
Writing Face Angle 45°
Color Fluorescent Yellow
Product Type Biopsy Cassette