missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ SH2D1A (Human) Recombinant Protein
Human SH2D1A full-length ORF ( AAH20732, 1 a.a. - 128 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00004068-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MDAVAVYHGKISRETGEKLLLATGLDGSYLLRDSESVPGVYCLCVLYHGYIYTYRVSQTETGSWSAETAPGVHKRYFRKIKNLISAFQKPDQGIVIPLQYPVEKKSSARSTQGTTGIREDPDVCLKAP
Specifications
AAH20732 | |
SH2 domain protein 1A | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
SH2D1A | |
GST |
4068 | |
Wheat germ expression system | |
25 μg | |
DSHP, EBVS, FLJ18687, FLJ92177, IMD5, LYP, MTCP1, SAP, XLP, XLPD | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |