missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ SGX 523

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
SGX 523 is a potent and selective c-MET kinase inhibitor (IC50 = 4nM). Exhibits minimal inhibition against a panel of 213 other protein kinases (at 1 μM). Suppresses growth of tumor xenografts derived from human glioblastoma, lung and gastric cancer cells.
Specifications
Specifications
| Quantity | 50 mg |
| Product Type | SGX 523 |
| Purity | >99% |
| Molecular Weight (g/mol) | 359.41 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction