Learn More
Trajan™ Thermo Scientific AS3000, AS2000 and AS200/800GC Autosampler Syringe
Product Code. 13425072
Click to view available options
Quantity:
1/Pk.
6/Pk.
Unit Size:
Each
Product Code. 13425072
Brand: Trajan™ 0029801
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Fixed needle autosampler syringes. For reduced non specific sample interaction and increased analyte recovery
  • High quality solutions for liquid handling
  • Accurate and reproducible injections
  • Specifically designed for Thermo Scientific instruments
Specifications
Needle Gauge 26 G
Diameter (Metric) Outer 0.47 mm
Product Type AS3000, AS2000 and AS200/800GC Autosampler Syringe
Quantity 1/Pk.
Volume (Metric) 10 μL