Learn More
SGE™ Series II Standard Fixed Needle Syringes
Product Code. 10518502
Change view
Click to view available options
Volume (Metric):
5 μL
10 μL
25 μL
50 μL
100 μL
250 μL
500 μL
Needle Gauge:
25 G
26 G
Includes:
Plunger protection
Standard plunger
Quantity:
10/Pk.
6 Pk.
Ea.
Unit Size:
Each
Pack of 10
Pack of 6
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Needle Gauge Includes Quantity unitSize
10518502 5 μL 26 G Plunger protection Ea. Each
12937536 10 μL 26 G Plunger protection 10/Pk. Pack of 10
10607171 10 μL 26 G Plunger protection Ea. Each
10127070 10 μL 26 G Plunger protection 6 Pk. Pack of 6
11941134 25 μL 25 G Standard plunger Ea. Each
10229010 50 μL 25 G Standard plunger Ea. Each
10625471 100 μL 25 G Standard plunger Ea. Each
10559072 250 μL 25 G Standard plunger Ea. Each
10771422 500 μL 25 G Standard plunger Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 10518502 Supplier SGE™ Supplier No. 001000
Save Up to 60% on Contract Testing Essentials
Use promo code 28025
View all promotions
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
Standard syringes with anti-roll flange and needles that are aligned with zero mark on barrel to reduce dead volume, improve accuracy and precision to <±1%
  • Stainless-steel plungers are hand fitted, noninterchangeable, nonreplaceable
  • Syringes with 5 and 10μL capacities have barrel O.D. of 6.5mm, scale length of 54.1mm and include a 50mm long 26s gauge needle
  • 25 to 500μL capacity syringes have 8mm O.D. barrel, 60mm scale length and 50mm long, 25 gauge needle Fixed-needle models have needle glued into barrel with solvent-resistant epoxy glue Removable-needle models feature a PTFE seal, a spring and nut for leak-resistant operation
Specifications
Accuracy ±1%
Material Borosilicate Glass (Barrel)
Needle Gauge 26 G
Diameter (English) Outer Needle 0.01 in.
Diameter (English) Outer 0.25 in.
Diameter (Metric) Outer 6.5 mm
Diameter (Metric) Outer Needle 0.47 mm
Includes Plunger protection
Length (English) Needle 1.96 in.
Product Type Microvolume Syringe
Graduations Major - 0.5 μL, Minor - 0.1 μL
Model 5F
Quantity Ea.
Volume (Metric) 5 μL
Needle Point Style Fixed Bevel
Connection Type Fixed