Learn More
Trajan™ Replacement Valves for Luer Lock Syringes
Product Code. 11947534
Click to view available options
For Use With (Equipment):
Syringe (5 mL to 2 L)
Syringe (50μL to 2 L)
Unit Size:
Each
Product Code. 11947534
Brand: Trajan™ undefined
To pressurize a sample prior to injection, a SGE syringe is pre-fitted with a valve
  • Push-pull valve with Luer Lock termination to suit removable needle 25μL-2.5mL syringes
  • Push button valve with Luer Lock termination. Suits 5mL-2L syringes with removable Luer Lock or removable needle
  • Luer Lock valve for any Luer Lock and Luer Tip syringes or other Luer devices. Ideal when fast needle changeover is needed
Specifications
Description Push buttone with Luer Lock
Product Type Replacement Valve
For Use With (Equipment) Syringe (5 mL to 2 L)