Learn More
Trajan™ Replacement Plungers for Diamond Manual Syringes
SGE syringe replacement plungers for gas tight syringes
Brand:  Trajan™ 0355105
Additional Details : Weight : 0.05000kg
 View more versions of this product
Product Code. 13187956
  • 64.90€ / Each
Estimated Shipment: 27-06-2024
Description
  • Replacement plungers for gas tight syringes
  • Designed to match the specific syringe barrel to maintain a gas tight seal
Specifications
Specifications
PTFE (Plunger)
Replacement Plunger
Diamond Manual Syringe (50μL)
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Sonderangebote
Sonderangebote