Learn More
Trajan™ Replacement Plungers for Diamond Manual Syringes
SGE syringe replacement plungers for gas tight syringes
64.90€
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
13167956
|
Trajan™
0355102 |
Diamond Manual Syringe (25μL) |
64.90€
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
13187956
|
Trajan™
0355105 |
Diamond Manual Syringe (50μL) |
64.90€
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
13107956
|
Trajan™
0355101 |
Diamond Manual Syringe (10μL) |
64.90€
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
13127966
|
Trajan™
0355110 |
Diamond Manual Syringe (100μL) |
64.90€
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
13137966
|
Trajan™
0355120 |
Diamond Manual Syringe (250μL) |
64.90€
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
13117956
|
Trajan™
0355150 |
Diamond Manual Syringe (500μL) |
64.90€
Each |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
Description
- Replacement plungers for gas tight syringes
- Designed to match the specific syringe barrel to maintain a gas tight seal
Specifications
PTFE (Plunger) |