Learn More
Trajan™ Replacement Plungers for Diamond Manual Syringes
SGE syringe replacement plungers for gas tight syringes
Brand: Trajan™ 0355110
Additional Details : Weight : 0.05000kg
Description
- Replacement plungers for gas tight syringes
- Designed to match the specific syringe barrel to maintain a gas tight seal
Specifications
PTFE (Plunger) | |
Replacement Plunger | |
Diamond Manual Syringe (100μL) |