Learn More
SGE™ MEPS™ Syringes
Product Code. 12967586
Change view
Click to view available options
Volume (Metric):
50 μL
100 μL
250 μL
500 μL
For Use With (Equipment):
CTC Analytics
CTC Analytics, HTA 300APlus, Thermo Scientific and Varian 8400 System
HTA 300APlus, Thermo Scientific and Varian 8400 System
eVol Automated Syringe
Product Type:
MEPS™ Syringe
Syringe
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) For Use With (Equipment) Product Type Unit Size
12967586 50 μL eVol Automated Syringe Syringe Each
11918516 250 μL HTA 300APlus, Thermo Scientific and Varian 8400 System MEPS™ Syringe Each
10296704 100 μL CTC Analytics, HTA 300APlus, Thermo Scientific and Varian 8400 System MEPS™ Syringe Each
11909068 250 μL CTC Analytics MEPS™ Syringe Each
12917596 500 μL eVol Automated Syringe Syringe Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 12967586 Supplier SGE™ Supplier No. 2910027
Estimated Shipment: 01-10-2026
Add to basket
Add to basket
For use in micro-SPE
  • The eVol™ custom programming function enables MEPS to be semi-automated and proofed before transition to fully automated platforms
Specifications
For Use With (Application) Gas/Liquid chromatography
Product Type Syringe
Quantity Ea.
Volume (Metric) 50 μL
For Use With (Equipment) eVol Automated Syringe