Learn More
Trajan™ HT8 GC Capillary Columns
Description
- High temperature, low bleed column
- Preferred column for polychlorinated biphenyl (PCB) analysis
- Separates PCBs on ortho ring as well as boiling point
Specifications
Specifications
| Max. Temperature | 360/370°C |
| Functionality/Interaction Type | End up with Pentavalent Bonds with Shared Sigma Bonds rather than piBond |
| Diameter (Metric) Inner | 0.32 mm |
| Stationary Phase | 8% Phenyl (equiv.) Polycarborane Siloxane |
| For Use With (Application) | Nitro-substituted Aromatics Analysis, PCB Congener Analysis, Nitro-substituted Aromatics, Polynuclear Aromatic Hydrocarbons, Pesticides and Herbicides Analysis |
| Temperature Range | -20°C to +360/370°C |
| Film Thickness | 0.25 μm |
| Length (Metric) | 25 m |
| Packing Material | Forte™ HT8 |
| Coating | Fused Silica |