Learn More
Trajan™ Diamond MS Thermo RSH and CTC RTC Autosampler Syringes
Click to view available options
Volume (Metric):
10 μL
25 μL
50 μL
100 μL
Model:
100R-RTC/RSH-GT-MS1
10R-RTC/RSH-GT-MS1
25R-RTC/RSH-GT-MS1
50R-RTC/RSH-GT-MS1
Unit Size:
Each
Description
- Lowest syringe carryover
- Reduced non-specific sample interaction and increased analyte recovery
- Compatible with ultra smooth removable needles with choice of surface treatment (sold separately)
Specifications
Specifications
| For Use With (Application) | Gas/Liquid chromatography |
| Product Type | Autosampler Syringe |
| Max. Temperature (English) | 662°F |
| Max. Temperature (Metric) | 350°C |
| Model | 50R-RTC/RSH-GT-MS1 |
| Quantity | Ea. |
| Volume (Metric) | 50 μL |
| For Use With (Equipment) | Thermo RSH and CTC RTC |
| Connection Type | Removable |
Product Content Correction
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur