missing translation for 'onlineSavingsMsg'
Learn More

Trajan™ Diamond LC Needles for 10 to 50μL Manual and Autosampler Syringes

Removable needle design ensures carryover levels are reduced even lower than the best fixed needle syringes available. Trajan Scientific and Medical™ Diamond LC Needles for 10 to 50μL Manual and Autosampler Syringes are MS ready and designed to improve the sensitivity of your MS results.

Brand:  Trajan™ 0355215

Additional Details : Weight : 0.05000kg

 View more versions of this product

Product Code. 13137906

  • 98.10€ / Each
Estimated Shipment: 28-06-2024
to see stock.
Add to basket

Description

Description

Improve the sensitivity of your MS analysis:

  • Lowest syringe carryover
  • Reduced non-specific sample interaction and increased analyte recovery
  • Ultra smooth removable needles with choice of surface treatment.
Specifications

Specifications

With Ultra Smooth Hydrophilic coating
0.028 in.
0.72 mm
Ultra smooth hydrophilic coating
Needle
LC
22 G
0.028 in.
0.72 mm
2.2 in.
57 mm
Manual and Autosampler Syringe (10 to 50μL)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers