Learn More
Trajan™ Diamond LC Needles for 10 to 50μL Manual and Autosampler Syringes
Removable needle design ensures carryover levels are reduced even lower than the best fixed needle syringes available. Trajan Scientific and Medical™ Diamond LC Needles for 10 to 50μL Manual and Autosampler Syringes are MS ready and designed to improve the sensitivity of your MS results.
90.40€ - 108.00€
Specifications
Needle Gauge | 22 G |
---|---|
Diameter (English) Outer | 0.028 in. |
Diameter (Metric) Outer | 0.72 mm |
Product Type | Needle |
Needle Point Style | LC |
Product Code | Brand | Diameter (Metric) Inner | Length (Metric) Needle | Description | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inner | Length (Metric) Needle | Description | Price | Quantity & Availability | |||
13107906
|
Trajan™
0355210 |
0.17 mm | 51 mm | With Ultra Smooth Hydrophilic coating |
98.10€
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||
13137906
|
Trajan™
0355215 |
0.72 mm | 57 mm | With Ultra Smooth Hydrophilic coating |
98.10€
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||
13157906
|
Trajan™
0355220 |
0.17 mm | 51 mm | With Ultra Smooth Hydrophobic coating |
108.00€
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||
13107916
|
Trajan™
0355230 |
0.17 mm | 51 mm | With Ultra Smooth coating |
90.40€
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
|||
13137916
|
Trajan™
0355235 |
0.17 mm | 57 mm | With Ultra Smooth coating |
90.40€
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||
13187906
|
Trajan™
0355225 |
0.17 mm | 57 mm | With Ultra Smooth Hydrophobic coating |
108.00€
Each |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||
Description
Improve the sensitivity of your MS analysis:
- Lowest syringe carryover
- Reduced non-specific sample interaction and increased analyte recovery
- Ultra smooth removable needles with choice of surface treatment.
Specifications
22 G | |
0.72 mm | |
LC |
0.028 in. | |
Needle | |
Manual and Autosampler Syringe (10 to 50μL) |